2'-Deoxyuridine ≥99%
Supplier: BeanTown Chemical
130775-1G
130775-25G
130775-5G
BT130775-1GEA
65.52
USD
BT130775-1G
BT130775-25G
BT130775-5G
2'-Deoxyuridine ≥99%
2'-Deoxyuridine
CAS: 951-78-0; EC No: 213-455-7; MDL No: MFCD00006527; RTECS: YU7490000
Crystalline; Molecular Formula: C9H12N2O5; MW: 228.20
Air Sensitive
- RTECS: YU7490000
- Air Sensitive
- Crystalline
Formula:
C₉H₁₂N₂O₅ MW: 228.20 g/mol |
MDL Number:
MFCD00006527 CAS Number: 951-78-0 EINECS: 213-455-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...