Linoleic acid ≥75%, Technical Grade

Supplier: BeanTown Chemical

130735-10G 130735-250G 130735-50G
BT130735-10GEA 54 USD
BT130735-10G BT130735-250G BT130735-50G
Linoleic acid ≥75%, Technical Grade
Linoleic acid

CAS: 60-33-3; EC No: 200-470-9; MDL No: MFCD00064241; RTECS: RF9990000 Liquid; Molecular Formula: C18H32O2; MW: 280.45 Melting Point: -5°; Boiling Point: 229-230°/16 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.902; Refractive Index: 1.466 Air Sensitive


  • RTECS: RF9990000
  • Refractive index: 1.466
  • Air Sensitive
  • Liquid

Formula: C₁₈H₃₂O₂
Boiling Pt: 229…230 °C/16 mmHg
Melting Pt: –5 °C
Density: 0.902 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9

Order Now

Specification Test Results

Assay 75%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR