4-Chloropyridine hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Chloropyridinium chloride

130675-100G 130675-25G 130675-5G
BT130675-100GEA 248 USD
BT130675-100G BT130675-25G BT130675-5G
4-Chloropyridine hydrochloride ≥98%
4-Chloropyridine hydrochloride

CAS: 7379-35-3; EC No: 230-946-1; MDL No: MFCD00012829; RTECS: US6300000 Solid; Molecular Formula: C5H4ClN·HCl ; MW: 150.01 Melting Point: 210° (sublimes); Flash point: 198°C (388°F)


  • RTECS: US6300000
  • Solid

Formula: C₅H₅Cl₂N
MW: 150.01 g/mol
Melting Pt: 210 °C (subl.)
Flash Pt: 198 °C (388 °F)
MDL Number: MFCD00012829
CAS Number: 7379-35-3
EINECS: 230-946-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR