Magnesium acetate tetrahydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate
130100-100G
130100-2.5KG
130100-500G
BT130100-100GEA
33.25
USD
BT130100-100G
BT130100-2.5KG
BT130100-500G
Magnesium acetate tetrahydrate 98.0-102.0% ACS
Magnesium acetate tetrahydrate
CAS: 16674-78-5; EC No: 205-554-9; MDL No: MFCD00149214
Crystalline; Linear Formula: (CH3COO)2Mg·4H2O; MW: 214.45
Melting Point: 72-75°
Density (g/mL): 1.454
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
(H₃CCOO)₂Mg·4H₂O Melting Pt: 72…75 °C Density: 1.454 g/mL |
MDL Number:
MFCD00149214 CAS Number: 16674-78-5 EINECS: 205-554-9 |
Specification Test Results
Assay | 98.0-102.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...