Bisacrylamide ≥97%

Supplier: BeanTown Chemical

Synonyms: N,N’-MethylenebisacrylamideN,N'-MethylenebisacrylamideBis (N,N'-methylenebisacrylamide)N,N-MethylenebisacrylamideN,N'-MethylenediacrylamideBIS

130085-500G 130085-100G
BT130085-100GEA 34.21 USD
BT130085-100G BT130085-500G
Bisacrylamide ≥97%
Bisacrylamide

  • RTECS: AS3678000
  • Air Sensitive, Light Sensitive
  • Solid

Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Melting Pt: >300 °C
Density: 1.235 g/mL
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://us.vwr.com/stibo/web/std.lang.all/70/49/2297049.jpg

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR