Benzocaine (ethyl-4-aminobenzoate) ≥98%

Supplier: BeanTown Chemical

130075-1KG 130075-250G 130075-5KG
BT130075-1KGEA 156.7 USD
BT130075-1KG BT130075-250G BT130075-5KG
Benzocaine (ethyl-4-aminobenzoate) ≥98%
Benzocaine (Ethyl 4-aminobenzoate)

CAS: 94-09-7; EC No: 202-303-5; MDL No: MFCD00007892; RTECS: DG2450000 Crystalline; Molecular Formula: C9H11NO2; MW: 165.19 Melting Point: 88-90°


  • RTECS: DG2450000
  • Crystalline

Formula: C₉H₁₁NO₂
Melting Pt: 88…90 °C
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR