6-Cyanonicotinic acid ≥97%

Supplier: BeanTown Chemical

130065-1G 130065-250MG
BT130065-1GEA 240 USD
BT130065-1G BT130065-250MG
6-Cyanonicotinic acid ≥97%
6-Cyanonicotinic acid

CAS: 70165-31-0; MDL No: MFCD01318135 Solid; Molecular Formula: C7H4N2O2; MW: 148.12 Melting Point: 190-194°


  • Solid

Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Melting Pt: 190…194 °C
MDL Number: MFCD01318135
CAS Number: 70165-31-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR