4-Bromo-3-chlorophenol ≥98%

Supplier: BeanTown Chemical

130050-1G 130050-5G
BT130050-1GEA 38 USD
BT130050-1G BT130050-5G
4-Bromo-3-chlorophenol ≥98%
4-Bromo-3-chlorophenol

CAS: 13631-21-5; MDL No: MFCD00070744 Crystalline/Powder; Molecular Formula: C6H4BrClO; MW: 207.46 Melting Point: 64-69°


  • Crystalline/Powder

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Melting Pt: 64…69 °C
MDL Number: MFCD00070744
CAS Number: 13631-21-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR