2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98%

Supplier: BeanTown Chemical

Synonyms: DavePhos2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl

129965-5G 129965-1G 129965-25G
BT129965-5GEA 383.32 USD
BT129965-5G BT129965-25G BT129965-1G
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl

  • Air Sensitive
  • Solid

Formula: C₂₆H₃₆NP
MW: 393.55 g/mol
Melting Pt: 121…124 °C
MDL Number: MFCD02183572
CAS Number: 213697-53-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR