Hydrochloric acid 1.0 N in water volumetric solution
Supplier: BeanTown Chemical
Synonyms:
Hydrochloric acid solution, Muriatic acid
129820-1L
129820-20L
129820-4L
BT129820-1LEA
48.98
USD
BT129820-1L
BT129820-20L
BT129820-4L
Hydrochloric acid 1.0 N in water volumetric solution
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324
Liquid; Molecular Formula: HCl; MW: 36.46
Density (g/mL): 1.017
- Liquid
Formula:
HCl Density: 1.017 g/mL |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...