Potassium sulfate 99,99

Supplier: BeanTown Chemical

129735-100G 129735-25G 129735-5G
BT129735-100GEA 401 USD
BT129735-100G BT129735-25G BT129735-5G
Potassium sulfate 99,99
Potassium sulphate

CAS: 7778-80-5; EC No: 231-915-5; MDL No: MFCD00011388; RTECS: TT5900000 Powder; Linear Formula: K2SO4; MW: 174.26 Melting Point: 1069°; Boiling Point: 1689° Density (g/mL): 2.662


  • RTECS: TT5900000
  • Powder

Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.662 g/mL
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR