2,5-Piperazinedione ≥99%

Supplier: BeanTown Chemical

Synonyms: Glycine anhydride, Piperazine-2,5-dione

129680-100G 129680-25G
BT129680-100GEA 156 USD
BT129680-100G BT129680-25G
2,5-Piperazinedione ≥99%
2,5-Piperazinedione

CAS: 106-57-0; EC No: 203-411-5; MDL No: MFCD00006009; RTECS: TL6325250 Powder; Molecular Formula: C4H6N2O2; MW: 114.10 Melting Point: 300°


  • RTECS: TL6325250
  • Powder

Formula: C₄H₆N₂O₂
MW: 114.1 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00006009
CAS Number: 106-57-0
EINECS: 203-411-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR