Calcium fluoride ≥99%
Supplier: BeanTown Chemical
129620-10G
129620-250G
129620-50G
BT129620-10GEA
48
USD
BT129620-10G
BT129620-250G
BT129620-50G
Calcium fluoride ≥99%
Calcium fluoride
CAS: 7789-75-5; EC No: 232-188-7; MDL No: MFCD00010907; RTECS: EW1760000
Crystalline; Molecular Formula: CaF2; MW: 78.08
Boiling Point: 2500°
Density (g/mL): 3.18
Hygroscopic
- RTECS: EW1760000
- Hygroscopic
- Crystalline
Formula:
CaF₂ Boiling Pt: 2500 °C Density: 3.18 g/mL |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...