Magnesium oxide ≥96%

Supplier: BeanTown Chemical

Synonyms: Magnesia usta

129555-100G 129555-500G
BT129555-100GEA 82.84 USD
BT129555-100G BT129555-500G
Magnesium oxide ≥96%
Magnesium oxide

CAS: 1309-48-4; EC No: 215-171-9; MDL No: MFCD00011109 -325 Mesh Powder; Molecular Formula: MgO ; MW: 40.30 Melting Point: 2852°; Boiling Point: 3600° Density (g/mL): 3.58; Refractive Index: 1.736 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.736
  • Air Sensitive, Moisture Sensitive
  • -325 Mesh Powder

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/mL
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR