Platinum ≥99.9% (trace metals basis), wire Ø 0.3 mm, hard type

Supplier: BeanTown Chemical

Synonyms: Platinium

128840-100CM 128840-25CM
BT128840-100CMEA 384.09 USD
BT128840-100CM BT128840-25CM
Platinum ≥99.9% (trace metals basis), wire Ø 0.3 mm, hard type
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 Solid; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Solid

Formula: Pt
Boiling Pt: 3827 °C
Melting Pt: 1772 °C
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR