trans-N-Boc-4-cyano-L-proline methyl ester ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-tert-Butyl 2-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate
128825-1G
128825-250MG
BT128825-1GEA
1161
USD
BT128825-1G
BT128825-250MG
trans-N-Boc-4-cyano-L-proline methyl ester ≥97%
trans-N-Boc-4-cyano-L-proline methyl ester
CAS: 194163-91-2; MDL No: MFCD03094915
Solid; Molecular Formula: C12H18N2O4; MW: 254.29
- Solid
Formula:
C₁₂H₁₈N₂O₄ MW: 254.286 g/mol |
MDL Number:
MFCD03094915 CAS Number: 194163-91-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...