Trimesic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid

128795-100G 128795-250G
BT128795-100GEA 91.87 USD
BT128795-100G BT128795-250G
Trimesic acid ≥98%
Trimesic acid

CAS: 554-95-0; EC No: 209-077-7; MDL No: MFCD00002517 Powder; Molecular Formula: C9H6O6; MW: 210.14


  • Powder

Formula: C₉H₆O₆
MDL Number: MFCD00002517
CAS Number: 554-95-0
EINECS: 209-077-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR