Vanadium ≥99.5% (trace metals basis), rods Ø 6.35 mm
Supplier: BeanTown Chemical
128785-100CM
128785-25CM
128785-5CM
BT128785-100CMEA
1750
USD
BT128785-100CM
BT128785-25CM
BT128785-5CM
Vanadium ≥99.5% (trace metals basis), rods Ø 6.35 mm
Vanadium
CAS: 7440-62-2; EC No: 231-171-1; MDL No: MFCD00011453; RTECS: YW1355000
Solid; Molecular Formula: V; MW: 50.94
Melting Point: 1890°; Boiling Point: 3380°
Density (g/mL): 6.11
- RTECS: YW1355000
- Solid
Formula:
V MW: 50.94 g/mol Boiling Pt: 3477 Melting Pt: 1896 Density: 6.11 g/mL |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 Merck Index: 12,10053 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...