n-Hexadecane ≥95%

Supplier: BeanTown Chemical

Synonyms: Hexadecane, Cetane

128425-1GAL 128425-5GAL
BT128425-1GALEA 179.34 USD
BT128425-1GAL BT128425-5GAL
n-Hexadecane ≥95%
n-Hexadecane

CAS: 544-76-3; EC No: 208-878-9; MDL No: MFCD00008998; RTECS: ML9200000 Liquid; Linear Formula: CH3(CH2)14CH3; Molecular Formula: C16H34; MW: 226.45 Melting Point: 18°; Boiling Point: 287°; Flash point: 135°C (275°F) Density (g/mL): 0.773; Refractive Index: 1.434 Hygroscopic


  • RTECS: ML9200000
  • Refractive index: 1.434
  • Hygroscopic
  • Liquid

Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
Boiling Pt: 287 °C
Melting Pt: 18 °C
Density: 0.773 g/mL
Flash Pt: 135 °C (275 °F)
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR