Tin ≥99.5% (trace metals basis), shot 8-20 mesh

Supplier: BeanTown Chemical

128415-100G 128415-2KG 128415-500G
BT128415-100GEA 49.81 USD
BT128415-100G BT128415-2KG BT128415-500G
Tin ≥99.5% (trace metals basis), shot 8-20 mesh
Tin

CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862; RTECS: XP7320000 Solid; Molecular Formula: Sn; MW: 118.71 Melting Point: 231.9°; Boiling Point: 2270° Density (g/mL): 7.31


  • RTECS: XP7320000
  • Solid

Formula: Sn
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.31 g/mL
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR