Chromium ≥99.99%, pieces 1-25 mm
Supplier: BeanTown Chemical
128090-1KG
128090-250G
128090-50G
BT128090-1KGEA
1513
USD
BT128090-1KG
BT128090-250G
BT128090-50G
Chromium ≥99.99%, pieces 1-25 mm
Chromium
CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944; RTECS: GB4200000
Broken Plate; Molecular Formula: Cr; MW: 52.00
Melting Point: 1857°; Boiling Point: 2672°
Density (g/mL): 7.14
- RTECS: GB4200000
- Broken Plate
Formula:
Cr MW: 52 Boiling Pt: 2469 Melting Pt: 1840 Density: 7.14 g/mL |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 Merck Index: 12,02288 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...