Lactic acid 1.0 N in water volumetric solution
Supplier: BeanTown Chemical
Synonyms:
DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
128050-1L
128050-500ML
BT128050-1LEA
103.28
USD
BT128050-1L
BT128050-500ML
Lactic acid 1.0 N in water volumetric solution
Lactic acid
CAS: 50-21-5; EC No: 200-018-0; MDL No: MFCD00004520
Liquid; Molecular Formula: C3H6O3 ; MW: 90.08
- Liquid
Formula:
H₃CCH(OH)COOH |
MDL Number:
MFCD00004520 CAS Number: 50-21-5 EINECS: 200-018-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...