Ammonium chloride 99,999

Supplier: BeanTown Chemical

Synonyms: Ammonium muriate, Salmiac

127865-1KG 127865-250G 127865-50G
BT127865-1KGEA 1213 USD
BT127865-1KG BT127865-250G BT127865-50G
Ammonium chloride 99,999
Ammonium chloride

CAS: 12125-02-9; EC No: 235-186-4; MDL No: MFCD00011420; RTECS: BP4550000 Powder; Linear Formula: NH4Cl; MW: 53.49 Melting Point: 340° (sublimes) Hygroscopic


  • RTECS: BP4550000
  • Hygroscopic
  • Powder

Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335 (subl.)
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR