Aluminium ≥99.999% (trace metals basis), foil, Thickness 0.025 mm

Supplier: BeanTown Chemical

Synonyms: Aluminum

127700-50X1000 127700-50X200
BT127700-50X10EA 341 USD
BT127700-50X10 BT127700-50X20
Aluminium ≥99.999% (trace metals basis), foil, Thickness 0.025 mm
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 Solid; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • RTECS: BD0330000
  • Solid

Formula: Al
Boiling Pt: 2460 °C
Melting Pt: 660.4 °C
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR