Silicon carbide, fine powder 320 grit
Supplier: BeanTown Chemical
127430-10KG
127430-2.5KG
127430-500G
BT127430-10KGEA
319.9
USD
BT127430-10KG
BT127430-2.5KG
BT127430-500G
Silicon carbide, fine powder 320 grit
Silicon carbide
CAS: 409-21-2; EC No: 206-991-8; MDL No: MFCD00049531
Powder; Linear Formula: SiC; MW: 40.10
Melting Point: 2700°
Density (g/mL): 3.22
- Powder
Formula:
SiC MW: 40.1 g/mol Melting Pt: 2700 °C Density: 3.22 g/mL |
MDL Number:
MFCD00049531 CAS Number: 409-21-2 EINECS: 206-991-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...