Methyl phenyl sulfoxide ≥98%

Supplier: BeanTown Chemical

Synonyms: (Methylsulphinyl)benzene, (Methylsulfinyl)benzene

127320-100G 127320-25G 127320-5G
BT127320-100GEA 485 USD
BT127320-100G BT127320-25G BT127320-5G
Methyl phenyl sulfoxide ≥98%
Methyl phenyl sulphoxide

CAS: 1193-82-4; EC No: 214-781-2; MDL No: MFCD00002088 Solid; Linear Formula: C6H5S(O)CH3; MW: 140.20 Melting Point: 26-29°; Boiling Point: 139-140°/14 mmHg; Flash point: 86°C (186°F) Refractive Index: 1.578 Hygroscopic


  • Refractive index: 1.578
  • Hygroscopic
  • Solid

Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 139…140 °C/14 mmHg
Melting Pt: 26…29 °C
Flash Pt: 86 °C (186 °F)
MDL Number: MFCD00002088
CAS Number: 1193-82-4
EINECS: 214-781-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR