D-(+)-Sucrose ACS
Supplier: BeanTown Chemical
127110-1KG
127110-250G
BT127110-1KGEA
87.25
USD
BT127110-1KG
BT127110-250G
D-(+)-Sucrose ACS
D(+)-Sucrose
CAS: 57-50-1; EC No: 200-334-9; MDL No: MFCD00006626; RTECS: WN6500000
Crystalline; Molecular Formula: C12H22O11; MW: 342.30
Melting Point: 185-187°
Density (g/mL): 1.58
- RTECS: WN6500000
- Crystalline
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 185…187 °C Density: 1.58 g/mL |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...