Zirconium(IV) oxide ≥99%

Supplier: BeanTown Chemical

Synonyms: Zirconium dioxide, Zirconium Oxide

127030-100G 127030-25G 127030-500G
BT127030-100GEA 230 USD
BT127030-100G BT127030-25G BT127030-500G
Zirconium(IV) oxide ≥99%
Zirconium (IV) oxide

CAS: 1314-23-4; EC No: 215-227-2; MDL No: MFCD00011310; RTECS: ZH8800000 15-25nm APS Powder, S.A. 20-30m 2/g; Linear Formula: ZrO2; MW: 123.22 Melting Point: 2700°; Boiling Point: 5000° Density (g/mL): 5.89


  • RTECS: ZH8800000
  • 15-25nm APS Powder, S.A. 20-30m 2/g

Formula: ZrO₂
Boiling Pt: 5000 °C
Melting Pt: 2700 °C
Density: 5.89 g/mL
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR