Folinic acid calcium salt ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt, Calcium (4-(((2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzoyl)-L-glutamate, Folinic acid calcium, Calcium folinate
127000-1G
127000-250MG
BT127000-1GEA
407
USD
BT127000-1G
BT127000-250MG
Folinic acid calcium salt ≥98%
Folinic acid calcium salt
CAS: 1492-18-8; EC No: 216-082-8; MDL No: MFCD00006704; RTECS: MA0600500
Solid; Molecular Formula: C20H21CaN7O7; MW: 511.50
- RTECS: MA0600500
Formula:
C₂₀H₂₁CaN₇O₇ MW: 511.51 g/mol |
MDL Number:
MFCD00006704 CAS Number: 1492-18-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...