6-Bromo-2-naphthoic acid ≥98%

Supplier: BeanTown Chemical

126960-25G 126960-5G
BT126960-25GEA 171 USD
BT126960-25G BT126960-5G
6-Bromo-2-naphthoic acid ≥98%
6-Bromo-2-naphthoic acid

CAS: 5773-80-8; MDL No: MFCD01075720 Powder; Molecular Formula: C11H7BrO2; MW: 251.08


  • Powder

Formula: C₁₁H₇BrO₂
MW: 251.08 g/mol
MDL Number: MFCD01075720
CAS Number: 5773-80-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR