Copper(II) acetylacetonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate
126955-100G
126955-25G
BT126955-100GEA
62.97
USD
BT126955-100G
BT126955-25G
Copper(II) acetylacetonate ≥98%
Copper (II) acetylacetonate
CAS: 13395-16-9; EC No: 236-477-9; MDL No: MFCD00000016; RTECS: GL6520000
Solid; Molecular Formula: C10H14CuO4 ; MW: 261.76
Melting Point: 284-288° (decomposes)
- RTECS: GL6520000
- Solid
Formula:
C₁₀H₁₄CuO₄ Melting Pt: 284…288 °C (dec.) |
MDL Number:
MFCD00000016 CAS Number: 13395-16-9 EINECS: 236-477-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...