6-Methoxy-2-methylpyridin-3-amine ≥95%

Supplier: BeanTown Chemical

Synonyms: 6-Methoxy-2-methylpyridin-3-amine, 3-Amino-6-methoxy-2-methylpyridine, 3-Amino-6-methoxy-2-picoline

126875-1G 126875-5G
BT126875-1GEA 112 USD
BT126875-1G BT126875-5G
6-Methoxy-2-methylpyridin-3-amine ≥95%
6-Methoxy-2-methylpyridin-3-amine

CAS: 52090-56-9 Solid; Molecular Formula: C7H10N20; MW: 138.17


  • Solid

Formula: C₇H₁₀N₂O
MW: 138.1671 g/mol
MDL Number: MFCD03095090
CAS Number: 52090-56-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR