Platinum ≥99.9% (trace metals basis), tubing 1.57 mm OD, 1.32 mm ID
Supplier: BeanTown Chemical
126825-25CM
126825-5CM
BT126825-25CMEA
1580
USD
BT126825-25CM
BT126825-5CM
Platinum ≥99.9% (trace metals basis), tubing 1.57 mm OD, 1.32 mm ID
Platinum
CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000
Solid; Molecular Formula: Pt; MW: 195.08
Melting Point: 1772°; Boiling Point: 3827°
Density (g/mL): 21.45
- RTECS: TP2160000
- Solid
Formula:
Pt Boiling Pt: 3827 °C Melting Pt: 1772 °C Density: 21.45 g/mL |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...