trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%

Supplier: BeanTown Chemical

126745-100G 126745-25G
BT126745-100GEA 199.48 USD
BT126745-100G BT126745-25G
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

CAS: 125572-95-4; EC No: 236-308-9; MDL No: MFCD00149243 Crystalline/Powder; Molecular Formula: C14H22N2O8·H2O ; MW: 364.35 Melting Point: 213-216°


  • Crystalline/Powder

Formula: C₁₄H₂₂N₂O₈·1H₂O
Melting Pt: 213…216 °C
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR