Aluminium ≥99.9995% (trace metals basis), foil, Thickness 1.0 µm

Supplier: BeanTown Chemical

Synonyms: Aluminum

126725-10MM 126725-25MM 126725-50MM
BT126725-10MMEA 624 USD
BT126725-10MM BT126725-25MM BT126725-50MM
Aluminium ≥99.9995% (trace metals basis), foil, Thickness 1.0 µm
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 Solid, on mylar support; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • RTECS: BD0330000
  • Solid, on mylar support

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

Specification Test Results

Assay 99.9995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR