Alizarin red S (sodium salt)
Supplier: BeanTown Chemical
Synonyms:
Alizarin sulfonic acid disodium salt, Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium
144735-25G
144735-5G
BT144735-25GEA
65.33
USD
BT144735-25G
BT144735-5G
Alizarin red S (sodium salt)
Alizarin Red S (sodium salt)
CAS: 130-22-3; EC No: 204-981-8; MDL No: MFCD00013049; RTECS: CB1095300
Powder; Molecular Formula: C14H7O7SNa; MW: 342.26
- RTECS: CB1095300
- Powder
Formula:
C₁₄H₇NaO₇S MW: 342.26 g/mol |
MDL Number:
MFCD00013049 CAS Number: 130-22-3 EINECS: 204-981-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...