Betaine, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
TMG, Trimethylglycine, 2-(Trimethylammonio)acetate
144585-100G
144585-2.5KG
144585-500G
BT144585-100GEA
46.39
USD
BT144585-100G
BT144585-2.5KG
BT144585-500G
Betaine, anhydrous ≥98%
Betaine
CAS: 107-43-7; EC No: 203-490-6; MDL No: MFCD00012123; RTECS: DS5900000
Powder; Linear Formula: (CH3)3NCH2CO2; Molecular Formula: C5H11NO2; MW: 117.15
Hygroscopic
- RTECS: DS5900000
- Hygroscopic
- Powder
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol |
MDL Number:
MFCD00012123 CAS Number: 107-43-7 EINECS: 203-490-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...