Trypan blue
Supplier: BeanTown Chemical
Synonyms:
Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate), AzidinBlue, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
CAS: 72-57-1; EC No: 200-786-7; MDL No: MFCD00003969; RTECS: QJ6475000
Solid; Molecular Formula: C34H24N6O14S4Na4; MW: 960.82
Melting Point:
- RTECS: QJ6475000
- Solid
Formula:
C₃₄H₂₄N₆Na₄O₁₄S₄ MW: 960.82 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00003969 CAS Number: 72-57-1 EINECS: 200-786-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...