2-bromo-5-methylthiazole ≥95%

Supplier: BeanTown Chemical

Synonyms: 2-Bromo-5-methylthiazole

126480-1G 126480-5G
BT126480-1GEA 63 USD
BT126480-1G BT126480-5G
2-bromo-5-methylthiazole ≥95%
2-bromo-5-methylthiazole

CAS: 41731-23-1 Solid; Molecular Formula: C4H4BrNS ; MW: 178.05


  • Solid

Formula: C₄H₄BrNS
MW: 178.047 g/mol
MDL Number: MFCD08460610
CAS Number: 41731-23-1

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR