Cadmium ≥99.95%, tear drop shot -5/+20 mesh

Supplier: BeanTown Chemical

126265-1KG 126265-250G 126265-50G
BT126265-1KGEA 293.42 USD
BT126265-1KG BT126265-250G BT126265-50G
Cadmium ≥99.95%, tear drop shot -5/+20 mesh
Cadmium

CAS: 7440-43-9; EC No: 231-152-8; MDL No: MFCD00010914 Solid; Molecular Formula: Cd; MW: 112.41 Melting Point: 320.9°; Boiling Point: 765° Density (g/mL): 8.65


  • Solid

Formula: Cd
Boiling Pt: 765 °C
Melting Pt: 320.9 °C
Density: 8.65 g/mL
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR