Tetraethylene glycol monomethyl ether ≥98%
Supplier: BeanTown Chemical
CAS: 23783-42-8; EC No: 245-883-5; MDL No: MFCD00041756
Liquid; Linear Formula: CH3(OCH2CH2)4OH; Molecular Formula: C9H20O5; MW: 208.25
Melting Point: -39°; Boiling Point: 163-166°/11 mmHg; Flash point:
- Refractive index: 1.444
- Liquid
Formula:
C₉H₂₀O₅ Boiling Pt: 163…166 °C/11 mmHg Melting Pt: –39 °C Density: 1.045 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00041756 CAS Number: 23783-42-8 EINECS: 245-883-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...