Silver(I) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Silver 2,4-pentanedionate, Silver acetylacetonate

125980-1G 125980-25G 125980-5G
BT125980-1GEA 78 USD
BT125980-1G BT125980-25G BT125980-5G
Silver(I) acetylacetonate
Silver(I) acetylacetonate

CAS: 15525-64-1; EC No: 239-568-1; MDL No: MFCD00000012 Solid; Linear Formula: [CH3COCH=C(O)CH3]Ag; Molecular Formula: C5H7AgO2; MW: 206.98 Melting Point: 100° (decomposes); Boiling Point: 225° Air Sensitive, Light Sensitive, Moisture Sensitive


  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Solid

Formula: C₅H₈AgO₂
Boiling Pt: 225 °C
Melting Pt: 100 °C (dec.)
MDL Number: MFCD00000012
CAS Number: 15525-64-1
EINECS: 239-568-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR