Ethylene carbonate ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,3-Dioxa-2-cyclopentanone

125730-10KG 125730-2.5KG 125730-500G
BT125730-10KGEA 310 USD
BT125730-10KG BT125730-2.5KG BT125730-500G
Ethylene carbonate ≥99%
Ethylene carbonate

CAS: 96-49-1; EC No: 202-510-0; MDL No: MFCD00005382; RTECS: FF9550000 Liquid/Low Melting Solid; Molecular Formula: C3H4O3; MW: 88.06 Melting Point: 35-38°; Boiling Point: 243-244°; Flash point: 143°C (289°F) Density (g/mL): 1.321


  • RTECS: FF9550000
  • Liquid/Low Melting Solid

Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 243
Melting Pt: 36
Density: 1.321 g/mL
Flash Pt: 143 °C (289 °F)
MDL Number: MFCD00005382
CAS Number: 96-49-1
EINECS: 202-510-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR