1,4-Butanesultone ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,4-Butane sultone

125590-100G 125590-25G
BT125590-100GEA 129.46 USD
BT125590-100G BT125590-25G
1,4-Butanesultone ≥99%
1,4-Butanesultone

CAS: 1633-83-6; EC No: 216-647-9; MDL No: MFCD00006584; RTECS: RP4300000 Liquid; Molecular Formula: C4H8O3S; MW: 136.17 Melting Point: 12-15°; Boiling Point: 165°/25 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.331; Refractive Index: 1.464 Moisture Sensitive


  • RTECS: RP4300000
  • Refractive index: 1.464
  • Moisture Sensitive
  • Liquid

Formula: C₄H₈O₃S
Boiling Pt: 165 °C/25 mmHg
Melting Pt: 12…15 °C
Density: 1.331 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00006584
CAS Number: 1633-83-6
EINECS: 216-647-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR