Sodium dihydrogen phosphate monohydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
125330-100G
125330-25G
BT125330-100GEA
375
USD
BT125330-100G
BT125330-25G
Sodium dihydrogen phosphate monohydrate ≥99.995% (trace metals basis)
Sodium dihydrogen phosphate monohydrate
CAS: 10049-21-5; EC No: 231-449-2; MDL No: MFCD00149208; RTECS: WA1900000
Crystalline; Linear Formula: NaH2PO4·H2O; MW: 137.99
Hygroscopic
- RTECS: WA1900000
- Hygroscopic
- Crystalline
Formula:
NaH₂PO₄·H₂O |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 EINECS: 231-449-2 |
|
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...