Diethanolamine ≥99%
Supplier: BeanTown Chemical
Synonyms:
ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol
125095-2.5KG
125095-500G
BT125095-2.5KGEA
91.29
USD
BT125095-2.5KG
BT125095-500G
Diethanolamine ≥99%
Diethanolamine
CAS: 111-42-2; EC No: 203-868-0; MDL No: MFCD00002843; RTECS: KL2975000
Liquid/Low Melting Solid; Linear Formula: HN(CH2CH2OH)2; Molecular Formula: C4H11NO2; MW: 105.14
Melting Point: 28°; Boiling Point: 217°/150 mmHg; Flash point: 138°C (280°F)
Density (g/mL): 1.097; Refractive Index: 1.477
Hygroscopic
- RTECS: KL2975000
- Refractive index: 1.477
- Hygroscopic
- Liquid/Low Melting Solid
Formula:
C₄H₁₁NO₂ MW: 105.14 g/mol Boiling Pt: 217 °C/150 mmHg Melting Pt: 28 °C Density: 1.097 g/mL Flash Pt: 138 °C (280 °F) |
MDL Number:
MFCD00002843 CAS Number: 111-42-2 EINECS: 203-868-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...