(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)oxazolidione

Supplier: BeanTown Chemical

Synonyms: (5R)-3-(3-Fluoro-4-(4-morpholinyl)phenyl)-5-hydroxymethyl-2-oxazolidione

124665-1G 124665-5G
BT124665-1GEA 234 USD
BT124665-1G BT124665-5G
(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)oxazolidione
(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)oxazolidione

CAS: 168828-82-8; MDL No: MFCD06658241 Solid; Molecular Formula: C14H17FN2O4; MW: 296.30


  • Solid

Formula: C₁₄H₁₇FN₂O₄
MW: 296.298 g/mol
MDL Number: MFCD06658241
CAS Number: 168828-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR