Vinpocetine

Supplier: BeanTown Chemical

Synonyms: (41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

124415-100MG 124415-20MG
BT124415-100MGEA 364 USD
BT124415-100MG BT124415-20MG
Vinpocetine
Vinpocetine

CAS: 42971-09-5; EC No: 256-028-0; MDL No: MFCD00211233; RTECS: JW4792000 Solid; Molecular Formula: C22H26N2O2; MW: 350.45 Melting Point: 147-149°


  • RTECS: JW4792000
  • Solid

Formula: C₂₂H₂₆N₂O₂
MW: 350.46 g/mol
Melting Pt: 147…149 °C
MDL Number: MFCD00211233
CAS Number: 42971-09-5
EINECS: 256-028-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR