Ammonium chloride ≥98%
Supplier: BeanTown Chemical
124085-1KG
124085-250G
124085-5KG
BT124085-1KGEA
40.89
USD
BT124085-1KG
BT124085-250G
BT124085-5KG
Ammonium chloride ≥98%
Ammonium chloride
CAS: 12125-02-9; EC No: 235-186-4; MDL No: MFCD00011420; RTECS: BP4550000
Solid; Linear Formula: NH4Cl; MW: 53.49
Melting Point: 340° (sublimes)
Hygroscopic
- RTECS: BP4550000
- Hygroscopic
- Solid
Formula:
NH₄Cl Melting Pt: 340 °C (subl.) |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...