Cyclohexylméthanol ≥99%

Supplier: BeanTown Chemical

124030-100G 124030-25G 124030-500G
BT124030-100GEA 94 USD
BT124030-100G BT124030-25G BT124030-500G
Cyclohexylméthanol ≥99%
Cyclohexylméthanol

CAS: 100-49-2; EC No: 202-857-8; MDL No: MFCD00001510; RTECS: GV5075000 Liquid; Molecular Formula: C7H14O; MW: 114.19 Boiling Point: 181°; Flash point: 77°C (171°F) Density (g/mL): 0.928; Refractive Index: 1.465


  • RTECS: GV5075000
  • Refractive index: 1.465
  • Liquid

Formula: C₇H₁₄O
Boiling Pt: 181 °C
Density: 0.928 g/mL
Flash Pt: 77 °C (171 °F)
MDL Number: MFCD00001510
CAS Number: 100-49-2
EINECS: 202-857-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR